Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACAAGCATGGAGGAAACAGACA[C/G]AACGACAGCGTTCAGGACAGTCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191720 | ||||||||||||||||||||
Literature Links: |
ARMC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC7 - armadillo repeat containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HN1 - hematological and neurological expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002032.2 | 778 | Missense Mutation | TCT,TGT | S,C 158 | NP_001002032.1 | |
NM_001002033.2 | 778 | UTR 3 | NP_001002033.1 | |||
NM_001288609.1 | 778 | UTR 3 | NP_001275538.1 | |||
NM_001288610.1 | 778 | UTR 3 | NP_001275539.1 | |||
NM_001288611.1 | 778 | UTR 3 | NP_001275540.1 | |||
NM_016185.3 | 778 | UTR 3 | NP_057269.1 |
NT5C - 5', 3'-nucleotidase, cytosolic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |