Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGCTGTGGGAGGTGTCAGTGG[C/T]GGACCGGCGCTCCCTCCTGGAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601964 MIM: 604497 | ||||||||||||||||||||
Literature Links: |
DNAJC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC7 - DnaJ heat shock protein family (Hsp40) member C7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKIRAS2 - NFKB inhibitor interacting Ras like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001349.2 | 520 | Missense Mutation | GCG,GTG | A,V 152 | NP_001001349.1 | |
NM_001144927.1 | 520 | Missense Mutation | GCG,GTG | A,V 152 | NP_001138399.1 | |
NM_001144928.1 | 520 | Missense Mutation | GCG,GTG | A,V 96 | NP_001138400.1 | |
NM_001144929.1 | 520 | UTR 3 | NP_001138401.1 | |||
NM_017595.5 | 520 | Missense Mutation | GCG,GTG | A,V 152 | NP_060065.2 | |
XM_005257251.1 | 520 | Missense Mutation | GCG,GTG | A,V 152 | XP_005257308.1 |
ZNF385C - zinc finger protein 385C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |