Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACTCACCGAGGCGCAGGGGCT[G/T]GAGGTGGTGCTGGGGGTAGGTGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606779 | ||||||||||||||||||||
Literature Links: |
EFCAB5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB5 - EF-hand calcium binding domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145053.1 | 98 | UTR 5 | NP_001138525.1 | |||
NM_198529.3 | 98 | Intron | NP_940931.2 | |||
XM_011524759.1 | 98 | Intron | XP_011523061.1 | |||
XM_011524763.2 | 98 | Intron | XP_011523065.1 | |||
XM_011524765.1 | 98 | Intron | XP_011523067.1 | |||
XM_011524766.2 | 98 | Intron | XP_011523068.1 | |||
XM_011524768.2 | 98 | Intron | XP_011523070.1 | |||
XM_017024591.1 | 98 | Intron | XP_016880080.1 | |||
XM_017024592.1 | 98 | Intron | XP_016880081.1 |