Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTTTCGGCAGGTTGTTAGTGCA[A/G]GCATCAATCTGGTGTATGGTAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605532 | ||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMURF2 - SMAD specific E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022739.3 | 2103 | Silent Mutation | GCC,GCT | A,A 705 | NP_073576.1 | |
XM_005257585.3 | 2103 | Silent Mutation | GCC,GCT | A,A 692 | XP_005257642.1 | |
XM_011525128.1 | 2103 | Silent Mutation | GCC,GCT | A,A 707 | XP_011523430.1 |