Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAGTAACAGGTCCTTGGGCCAG[C/T]GCCAGAACTCTCCGCTGCCCTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610097 | ||||||||||||||||||||
Literature Links: |
ODF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ODF4 - outer dense fiber of sperm tails 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319953.1 | 363 | Intron | NP_001306882.1 | |||
NM_153007.4 | 363 | Missense Mutation | CGC,TGC | R,C 61 | NP_694552.2 | |
XM_005256476.3 | 363 | Intron | XP_005256533.1 | |||
XM_011523688.1 | 363 | Intron | XP_011521990.1 |