Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCGGGCTGGAGAACCGGCGAG[A/C]CCCCTGAGGATGCAGACAAACATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614271 MIM: 180073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC137 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC137 - coiled-coil domain containing 137 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OXLD1 - oxidoreductase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039842.2 | 124 | Missense Mutation | GCT,TCT | A,S 22 | NP_001034931.1 | |
NM_001304994.1 | 124 | Missense Mutation | GCT,TCT | A,S 12 | NP_001291923.1 | |
NM_001304995.1 | 124 | UTR 3 | NP_001291924.1 | |||
NM_001304999.1 | 124 | UTR 3 | NP_001291928.1 |
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |