Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCACTGTCACACAGATGCACTT[C/T]TTGACCGGCCAGGTGAGCCTCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600966 | ||||||||||||||||||||
Literature Links: |
LLGL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LLGL1 - LLGL1, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004140.3 | 310 | Silent Mutation | TTC,TTT | F,F 83 | NP_004131.3 | |
XM_011523849.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522151.1 | |
XM_011523850.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522152.1 | |
XM_011523851.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522153.1 | |
XM_011523852.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522154.1 | |
XM_011523853.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522155.1 | |
XM_011523854.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522156.1 | |
XM_011523855.2 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522157.1 | |
XM_011523856.1 | 310 | Silent Mutation | TTC,TTT | F,F 83 | XP_011522158.1 |