Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCTTTCTTCCCTGCTCCTTTGC[A/C]TTCCAGGACCTCAGCCACCCTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613212 | ||||||||||||||||||||
Literature Links: |
AARSD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AARSD1 - alanyl-tRNA synthetase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261434.1 | 1991 | Missense Mutation | NP_001248363.1 |
PTGES3L-AARSD1 - PTGES3L-AARSD1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136042.2 | 1991 | Missense Mutation | NP_001129514.2 | |||
NM_025267.3 | 1991 | Missense Mutation | NP_079543.1 |