Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACTCGATGCCTTCGTATTTCAC[A/G]CTTCCGATCTTGGTCTCCGGCAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613165 | ||||||||||||||||||||
Literature Links: |
CANT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CANT1 - calcium activated nucleotidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159772.1 | 1594 | Silent Mutation | NP_001153244.1 | |||
NM_001159773.1 | 1594 | Silent Mutation | NP_001153245.1 | |||
NM_138793.3 | 1594 | Silent Mutation | NP_620148.1 | |||
XM_005257021.1 | 1594 | Silent Mutation | XP_005257078.1 | |||
XM_005257022.1 | 1594 | Silent Mutation | XP_005257079.1 | |||
XM_006721683.1 | 1594 | Silent Mutation | XP_006721746.1 | |||
XM_011524291.1 | 1594 | Silent Mutation | XP_011522593.1 | |||
XM_011524293.1 | 1594 | Silent Mutation | XP_011522595.1 | |||
XM_011524294.1 | 1594 | Silent Mutation | XP_011522596.1 | |||
XM_011524295.2 | 1594 | Silent Mutation | XP_011522597.1 |