Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131370 MIM: 606672 MIM: 176610 MIM: 610431 MIM: 604165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENO3 - enolase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GP1BA - glycoprotein Ib platelet alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFN1 - profilin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF167 - ring finger protein 167 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320356.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307285.1 | |
NM_001320357.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307286.1 | |
NM_001320358.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307287.1 | |
NM_001320359.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307288.1 | |
NM_001320360.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307289.1 | |
NM_001320361.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_001307290.1 | |
NM_001320362.1 | 515 | Silent Mutation | GCA,GCG | A,A 50 | NP_001307291.1 | |
NM_001320363.1 | 515 | Silent Mutation | GCA,GCG | A,A 50 | NP_001307292.1 | |
NM_001320364.1 | 515 | Silent Mutation | GCA,GCG | A,A 50 | NP_001307293.1 | |
NM_001320365.1 | 515 | Silent Mutation | GCA,GCG | A,A 50 | NP_001307294.1 | |
NM_015528.2 | 515 | Silent Mutation | GCA,GCG | A,A 85 | NP_056343.1 | |
XM_017024427.1 | 515 | Silent Mutation | GCA,GCG | A,A 85 | XP_016879916.1 | |
XM_017024428.1 | 515 | Intron | XP_016879917.1 |
SLC25A11 - solute carrier family 25 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |