Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGCTCGCTTCTTGCAGCAGCC[A/G]CGGCACAGGCTGAACACACATCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601934 MIM: 602578 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUS1L - dihydrouridine synthase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022156.4 | 1398 | Silent Mutation | CGC,CGT | R,R 412 | NP_071439.3 | |
XM_005256393.2 | 1398 | Missense Mutation | CGG,TGG | R,W 534 | XP_005256450.1 | |
XM_005256394.2 | 1398 | Missense Mutation | CGG,TGG | R,W 410 | XP_005256451.1 | |
XM_006722288.2 | 1398 | Missense Mutation | CGG,TGG | R,W 517 | XP_006722351.1 | |
XM_006722289.2 | 1398 | Silent Mutation | CGC,CGT | R,R 395 | XP_006722352.1 |
RFNG - RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |