Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGCCATCTCCTTCAACTTGC[A/G]CTTCACCAGTGCCCGGTCATGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602525 | ||||||||||||||||||||
Literature Links: |
PDK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDK2 - pyruvate dehydrogenase kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAMD14 - sterile alpha motif domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257359.1 | 2458 | Missense Mutation | CGC,TGC | R,C 381 | NP_001244288.1 | |
NM_174920.3 | 2458 | Missense Mutation | CGC,TGC | R,C 409 | NP_777580.1 | |
XM_011524490.2 | 2458 | Missense Mutation | CGC,TGC | R,C 164 | XP_011522792.1 | |
XM_017024322.1 | 2458 | Missense Mutation | CGC,TGC | R,C 464 | XP_016879811.1 | |
XM_017024323.1 | 2458 | Missense Mutation | CGC,TGC | R,C 381 | XP_016879812.1 | |
XM_017024324.1 | 2458 | Missense Mutation | CGC,TGC | R,C 409 | XP_016879813.1 |