Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGGGACAATTAAATAAAATGA[A/G]GAAGGAAAATAATACTATTTGCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRC3 - dynein regulatory complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130090.1 | Intron | NP_001123562.1 | ||||
NM_001130091.1 | Intron | NP_001123563.1 | ||||
NM_001130092.1 | Intron | NP_001123564.1 | ||||
NM_031294.3 | Intron | NP_112584.3 | ||||
XM_006721583.3 | Intron | XP_006721646.1 | ||||
XM_011524018.2 | Intron | XP_011522320.1 | ||||
XM_011524019.2 | Intron | XP_011522321.1 | ||||
XM_011524020.2 | Intron | XP_011522322.1 | ||||
XM_011524021.2 | Intron | XP_011522323.1 | ||||
XM_011524022.2 | Intron | XP_011522324.1 | ||||
XM_011524023.2 | Intron | XP_011522325.1 | ||||
XM_011524025.2 | Intron | XP_011522327.1 | ||||
XM_011524026.2 | Intron | XP_011522328.1 |
TOM1L2 - target of myb1 like 2 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |