Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCGCCTGGTGAATGGGCTGAGT[A/G]GGCCAAGTGGGTGGGCTGGGGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160993 MIM: 608795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6784 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6784 - microRNA 6784 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMT1 - N-myristoyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCD3 - phospholipase C delta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133373.4 | 2612 | UTR 3 | NP_588614.1 | |||
XM_011524253.2 | 2612 | Intron | XP_011522555.1 |