Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCTTTCCAGCACCTTCACTCG[G/T]CCTGTTTTTCTTCCTCCTCCTCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142966 MIM: 142960 MIM: 142961 | ||||||||||||||||||||
Literature Links: |
HOXB-AS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXB-AS3 - HOXB cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB3 - homeobox B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB5 - homeobox B5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB6 - homeobox B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018952.4 | 1235 | Missense Mutation | GAC,GCC | D,A 223 | NP_061825.2 | |
XM_005257283.3 | 1235 | Missense Mutation | GAC,GCC | D,A 223 | XP_005257340.1 | |
XM_005257284.3 | 1235 | Missense Mutation | GAC,GCC | D,A 223 | XP_005257341.1 | |
XM_011524727.2 | 1235 | Missense Mutation | GAC,GCC | D,A 287 | XP_011523029.1 |