Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAGGAGAGTAACAACTCACCGA[C/T]TTCACTCGGTGACCAATGCCCATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108728 | ||||||||||||||||||||
Literature Links: |
ACLY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACLY - ATP citrate lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001096.2 | 3104 | Silent Mutation | AAA,AAG | K,K 978 | NP_001087.2 | |
NM_001303274.1 | 3104 | Silent Mutation | AAA,AAG | K,K 1032 | NP_001290203.1 | |
NM_001303275.1 | 3104 | Silent Mutation | AAA,AAG | K,K 1022 | NP_001290204.1 | |
NM_198830.1 | 3104 | Silent Mutation | AAA,AAG | K,K 968 | NP_942127.1 | |
XM_005257395.1 | 3104 | Silent Mutation | AAA,AAG | K,K 978 | XP_005257452.1 | |
XM_017024688.1 | 3104 | Silent Mutation | AAA,AAG | K,K 968 | XP_016880177.1 |
KLHL11 - kelch like family member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |