Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCCGAGTGTGGGAATCGGCTCG[C/G]GAGTGCGGGCTTCGGGGCTCTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 111730 | ||||||||||||||||||||
Literature Links: |
B4GALNT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B4GALNT2 - beta-1,4-N-acetyl-galactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159387.1 | 313 | Intron | NP_001152859.1 | |||
NM_001159388.1 | 313 | Intron | NP_001152860.1 | |||
NM_153446.2 | 313 | Missense Mutation | GCG,GGG | A,G 34 | NP_703147.2 | |
XM_011524314.2 | 313 | Missense Mutation | GCG,GGG | A,G 34 | XP_011522616.1 | |
XM_017024173.1 | 313 | Intron | XP_016879662.1 |