Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACTGAAGAAAGAATCTGCATGT[G/T]TGAGACTATTCAAGCAAGAAGGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602627 MIM: 609692 | ||||||||||||||||||||
Literature Links: |
CDC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC6 - cell division cycle 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254.3 | 854 | Missense Mutation | GTG,TTG | V,L 147 | NP_001245.1 | |
XM_011525541.1 | 854 | Missense Mutation | GTG,TTG | V,L 147 | XP_011523843.1 | |
XM_011525542.1 | 854 | Missense Mutation | GTG,TTG | V,L 147 | XP_011523844.1 |
WIPF2 - WAS/WASL interacting protein family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |