Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCACCAACATCGTGTCAGTGTGC[C/T]GCGCCACTGGGCTGGGGCCTGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613129 MIM: 602133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTC1 - CTS telomere maintenance complex component 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFAS - phosphoribosylformylglycinamidine synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012393.2 | 466 | Missense Mutation | CGC,TGC | R,C 109 | NP_036525.1 | |
XM_006721546.2 | 466 | Missense Mutation | CGC,TGC | R,C 109 | XP_006721609.1 | |
XM_017024760.1 | 466 | UTR 5 | XP_016880249.1 |