Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCATCTTAGAAAGTTCTCGAAA[C/T]GCTTCATCCCCGACAAAGCAAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GLOD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLOD4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016080.3 | 878 | Silent Mutation | GCA,GCG | A,A 260 | NP_057164.3 | |
XM_006721540.2 | 878 | Silent Mutation | GCA,GCG | A,A 251 | XP_006721603.1 | |
XM_017024711.1 | 878 | Silent Mutation | GCA,GCG | A,A 251 | XP_016880200.1 | |
XM_017024712.1 | 878 | Silent Mutation | GCA,GCG | A,A 204 | XP_016880201.1 |