Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCTGACACAGTACTTGAATTC[C/T]AAAAAGTCTATGGTGTCTTCACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEATR6 - HEAT repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022070.4 | 5134 | Silent Mutation | TTA,TTG | L,L 1060 | NP_071353.4 | |
XM_005257570.3 | 5134 | Silent Mutation | TTA,TTG | L,L 892 | XP_005257627.1 | |
XM_011525110.2 | 5134 | Silent Mutation | TTA,TTG | L,L 1024 | XP_011523412.1 | |
XM_011525111.2 | 5134 | Silent Mutation | TTA,TTG | L,L 1013 | XP_011523413.1 | |
XM_017024936.1 | 5134 | Silent Mutation | TTA,TTG | L,L 892 | XP_016880425.1 | |
XM_017024937.1 | 5134 | Silent Mutation | TTA,TTG | L,L 892 | XP_016880426.1 | |
XM_017024938.1 | 5134 | Silent Mutation | TTA,TTG | L,L 599 | XP_016880427.1 | |
XM_017024939.1 | 5134 | Silent Mutation | TTA,TTG | L,L 599 | XP_016880428.1 |
MIR4737 - microRNA 4737 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |