Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGGCACTACGCCACGCAGCT[C/T]CAGGTGGGTGTAGGCTCTGGGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAT5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STAT5A - signal transducer and activator of transcription 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288718.1 | 542 | Silent Mutation | CTC,CTT | L,L 94 | NP_001275647.1 | |
NM_001288719.1 | 542 | Silent Mutation | CTC,CTT | L,L 94 | NP_001275648.1 | |
NM_001288720.1 | 542 | Silent Mutation | CTC,CTT | L,L 94 | NP_001275649.1 | |
NM_003152.3 | 542 | Silent Mutation | CTC,CTT | L,L 94 | NP_003143.2 | |
XM_005257624.3 | 542 | Silent Mutation | CTC,CTT | L,L 94 | XP_005257681.1 |