Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTCCTTCAAGGTGCAGCTGGC[A/G]CCGGCCAGATCGGCTGGAAGAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160776 MIM: 607538 | ||||||||||||||||||||
Literature Links: |
MYH10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYH10 - myosin, heavy chain 10, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256012.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1975 | NP_001242941.1 | |
NM_001256095.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1953 | NP_001243024.1 | |
NM_005964.3 | 6077 | Missense Mutation | CGC,TGC | R,C 1944 | NP_005955.3 | |
XM_011523875.2 | 6077 | Missense Mutation | CGC,TGC | R,C 2005 | XP_011522177.1 | |
XM_011523878.2 | 6077 | Missense Mutation | CGC,TGC | R,C 1984 | XP_011522180.1 | |
XM_011523879.2 | 6077 | Missense Mutation | CGC,TGC | R,C 1983 | XP_011522181.1 | |
XM_011523880.2 | 6077 | Missense Mutation | CGC,TGC | R,C 1974 | XP_011522182.1 | |
XM_017024677.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1954 | XP_016880166.1 | |
XM_017024678.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1954 | XP_016880167.1 | |
XM_017024679.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1954 | XP_016880168.1 | |
XM_017024680.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1953 | XP_016880169.1 | |
XM_017024681.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1944 | XP_016880170.1 | |
XM_017024682.1 | 6077 | Missense Mutation | CGC,TGC | R,C 1944 | XP_016880171.1 |
NDEL1 - nudE neurodevelopment protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |