Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATCACTAAAAATGACCCACCTG[C/T]TCCAGGGTAGACTGTGAGAGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612507 | ||||||||||||||||||||
Literature Links: |
ABCA9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA9 - ATP binding cassette subfamily A member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080283.3 | 4881 | Silent Mutation | GAA,GAG | E,E 1591 | NP_525022.2 | |
XM_017024011.1 | 4881 | Silent Mutation | GAA,GAG | E,E 1599 | XP_016879500.1 | |
XM_017024012.1 | 4881 | Silent Mutation | GAA,GAG | E,E 1591 | XP_016879501.1 | |
XM_017024013.1 | 4881 | Silent Mutation | GAA,GAG | E,E 1591 | XP_016879502.1 | |
XM_017024014.1 | 4881 | Silent Mutation | GAA,GAG | E,E 1591 | XP_016879503.1 | |
XM_017024015.1 | 4881 | Silent Mutation | GAA,GAG | E,E 1441 | XP_016879504.1 |