Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCATTCGTGACCTTTCTCTGC[C/T]CCCTGTGGATCGACTTGGATTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606793 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPEPPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPEPPS - aminopeptidase puromycin sensitive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006310.3 | 2017 | Missense Mutation | CCC,CTC | P,L 611 | NP_006301.3 | |
XM_011525496.1 | 2017 | Missense Mutation | CCC,CTC | P,L 607 | XP_011523798.1 | |
XM_017025372.1 | 2017 | Missense Mutation | CCC,CTC | P,L 611 | XP_016880861.1 | |
XM_017025373.1 | 2017 | Missense Mutation | CCC,CTC | P,L 607 | XP_016880862.1 |