Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAATTCAAGTCTCCCAAGCGGT[C/T]GGCTAGGGACCGAATGGAGAAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604260 | ||||||||||||||||||||
Literature Links: |
STAT5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STAT5B - signal transducer and activator of transcription 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012448.3 | 1960 | Missense Mutation | AAC,GAC | N,D 658 | NP_036580.2 | |
XM_005257626.4 | 1960 | Intron | XP_005257683.1 | |||
XM_017024977.1 | 1960 | Missense Mutation | AAC,GAC | N,D 564 | XP_016880466.1 |