Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAACCTTTTCTTAATTCCATTCC[A/G]TAGTCCTCAACAAGTGTGGACCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604450 | ||||||||||||||||||||
Literature Links: |
PSMD12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMD12 - proteasome 26S subunit, non-ATPase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316341.1 | 1364 | Silent Mutation | TAC,TAT | Y,Y 265 | NP_001303270.1 | |
NM_002816.4 | 1364 | Silent Mutation | TAC,TAT | Y,Y 324 | NP_002807.1 | |
NM_174871.3 | 1364 | Silent Mutation | TAC,TAT | Y,Y 304 | NP_777360.1 |