Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGTTCCGAAGTCGGCGAGGGA[A/G]GGGGTTTACTTGCCCGATCGTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604607 MIM: 609819 MIM: 610787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXB13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXB13 - homeobox B13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006361.5 | 2751 | UTR 3 | NP_006352.2 |
MIR3185 - microRNA 3185 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAC1 - prostate cancer susceptibility candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAC2 - prostate cancer susceptibility candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282275.1 | 2751 | Intron | NP_001269204.1 | |||
NM_001282276.1 | 2751 | Intron | NP_001269205.1 | |||
XM_006721865.3 | 2751 | Intron | XP_006721928.1 | |||
XM_011524747.2 | 2751 | Intron | XP_011523049.1 | |||
XM_011524748.1 | 2751 | Intron | XP_011523050.1 |