Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGTTGATGAAATCCACTGCCTG[A/G]GTTTTCAACTGATCTGCACTGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602650 | ||||||||||||||||||||
Literature Links: |
SPOP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPOP - speckle type BTB/POZ protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007226.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_001007227.1 | |
NM_001007227.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_001007228.1 | |
NM_001007228.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_001007229.1 | |
NM_001007229.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_001007230.1 | |
NM_001007230.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_001007231.1 | |
NM_003563.3 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | NP_003554.1 | |
XM_005257723.4 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | XP_005257780.1 | |
XM_005257724.4 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | XP_005257781.1 | |
XM_017025204.1 | 1428 | Silent Mutation | ACC,ACT | T,T 319 | XP_016880693.1 |