Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTTCTGCATCTAGTAAGTCCAG[A/G]AGCAAGTGGAACACTTGTGGACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEATR9 - HEAT repeat containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321395.1 | 1412 | Silent Mutation | CTC,CTT | L,L 400 | NP_001308324.1 | |
NM_152781.3 | 1412 | Silent Mutation | CTC,CTT | L,L 440 | NP_689994.2 | |
XM_005278233.2 | 1412 | Silent Mutation | CTC,CTT | L,L 406 | XP_005278290.1 | |
XM_005278235.1 | 1412 | Silent Mutation | CTC,CTT | L,L 366 | XP_005278292.1 | |
XM_006721802.3 | 1412 | Silent Mutation | CTC,CTT | L,L 258 | XP_006721865.1 | |
XM_011524586.2 | 1412 | Silent Mutation | CTC,CTT | L,L 406 | XP_011522888.1 | |
XM_017024424.1 | 1412 | Silent Mutation | CTC,CTT | L,L 440 | XP_016879913.1 |
TAF15 - TATA-box binding protein associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |