Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGGTTAGGGGGGTTCTGGGGC[A/G]TTGGAGTCCGACTGGCCTTGGGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607217 | ||||||||||||||||||||
Literature Links: |
SDK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDK2 - sidekick cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144952.1 | 6407 | Missense Mutation | ACG,ATG | T,M 2136 | NP_001138424.1 | |
XM_011524914.2 | 6407 | Missense Mutation | ACG,ATG | T,M 2117 | XP_011523216.1 | |
XM_011524915.2 | 6407 | Intron | XP_011523217.1 | |||
XM_011524916.2 | 6407 | Intron | XP_011523218.1 |