Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCGTCTCCCGGGCTGTGAGCC[A/G]CCCTGCTGTCCTGAGTGCCGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF112 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF112 - ring finger protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007148.4 | 245 | Silent Mutation | CCA,CCG | P,P 91 | NP_009079.2 | |
XM_006721571.3 | 245 | Silent Mutation | CCA,CCG | P,P 91 | XP_006721634.1 | |
XM_006721572.3 | 245 | Silent Mutation | CCA,CCG | P,P 91 | XP_006721635.1 | |
XM_006721573.3 | 245 | Silent Mutation | CCA,CCG | P,P 67 | XP_006721636.1 |