Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCACCTGGGAGAGGTTGTAGTCCG[C/T]GCCCAGGTCCTCGCCGTAGGCGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASN - fatty acid synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004104.4 | 7495 | Missense Mutation | ACG,GCG | T,A 2460 | NP_004095.4 | |
XM_011523538.2 | 7495 | Missense Mutation | ACG,GCG | T,A 2460 | XP_011521840.1 |