Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCAGGGTGTCCACCGGAGAAG[A/C]CTTCGGGGCTGGCACATGGGACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603697 | ||||||||||||||||||||
Literature Links: |
ALOX15B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALOX15B - arachidonate 15-lipoxygenase, type B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039130.1 | 154 | Missense Mutation | GAC,GCC | D,A 13 | NP_001034219.1 | |
NM_001039131.1 | 154 | Missense Mutation | GAC,GCC | D,A 13 | NP_001034220.1 | |
NM_001141.2 | 154 | Missense Mutation | GAC,GCC | D,A 13 | NP_001132.2 |