Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATAATTTTTGCCACACCTGTG[C/G]TCCCCACATTTTTTGACTCTGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609086 MIM: 604311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL20 - F-box and leucine rich repeat protein 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED1 - mediator complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004774.3 | 4302 | Missense Mutation | ACC,AGC | T,S 1386 | NP_004765.2 | |
XM_005257465.3 | 4302 | Missense Mutation | ACC,AGC | T,S 1214 | XP_005257522.1 | |
XM_006721957.1 | 4302 | Intron | XP_006722020.1 | |||
XM_017024779.1 | 4302 | Missense Mutation | ACC,AGC | T,S 1167 | XP_016880268.1 |