Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTGCCCCAACAGGCCCTCCAC[A/G]CCCTTCAGGAGGAGCAGGCCAGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600735 MIM: 607526 MIM: 604631 | ||||||||||||||||||||
Literature Links: |
IFI35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFI35 - interferon induced protein 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005533.4 | 239 | Missense Mutation | ACC,GCC | T,A 11 | NP_005524.2 | |
XM_005257302.4 | 239 | Missense Mutation | ACC,GCC | T,A 11 | XP_005257359.1 | |
XM_017024584.1 | 239 | UTR 5 | XP_016880073.1 | |||
XM_017024585.1 | 239 | UTR 5 | XP_016880074.1 |
RPL27 - ribosomal protein L27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAT1 - vesicle amine transport 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |