Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTAGCCCGCGGGATGAGGATG[A/G]CACCAAGGCAGACAGCTGGAAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600362 MIM: 615498 MIM: 601243 | ||||||||||||||||||||
Literature Links: |
FLII PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLII - FLII, actin remodeling protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIEF2 - mitochondrial elongation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144900.1 | 396 | Missense Mutation | GAC,GGC | D,G 62 | NP_001138372.1 | |
NM_139162.3 | 396 | Missense Mutation | GAC,GGC | D,G 62 | NP_631901.2 | |
NM_148886.1 | 396 | Missense Mutation | GAC,GGC | D,G 73 | NP_683684.2 | |
XM_017024190.1 | 396 | Missense Mutation | GAC,GGC | D,G 69 | XP_016879679.1 |
TOP3A - topoisomerase (DNA) III alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |