Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTCTAGTTTTGTCACAAAGGCT[C/T]ATGCTGTTCCGTGTCCTCTTCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG1 - component of oligomeric golgi complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018714.2 | 1944 | Intron | NP_061184.1 |
FAM104A - family with sequence similarity 104 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098832.1 | 1944 | UTR 3 | NP_001092302.1 | |||
NM_001289410.1 | 1944 | UTR 3 | NP_001276339.1 | |||
NM_001289411.1 | 1944 | UTR 3 | NP_001276340.1 | |||
NM_001289412.1 | 1944 | UTR 3 | NP_001276341.1 | |||
NM_032837.2 | 1944 | UTR 3 | NP_116226.2 |