Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTCCGAAATGCTGCAATCGCCT[C/T]TTCTGGGCCTGGGGGAGGAAGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607130 | ||||||||||||||||||||
Literature Links: |
RPTOR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPTOR - regulatory associated protein of MTOR complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163034.1 | 833 | Missense Mutation | CTT,TTT | L,F 10 | NP_001156506.1 | |
NM_020761.2 | 833 | Missense Mutation | CTT,TTT | L,F 10 | NP_065812.1 |