Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCGTCATCGTCCATGCCCCCAAA[C/T]ACAGTAATGACATCCCCTGCCCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610764 | ||||||||||||||||||||
Literature Links: |
TSPOAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPOAP1 - TSPO associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261835.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1791 | NP_001248764.1 | |
NM_004758.3 | 5255 | Silent Mutation | GTA,GTG | V,V 1800 | NP_004749.2 | |
NM_024418.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1740 | NP_077729.1 | |
XM_005257807.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1791 | XP_005257864.1 | |
XM_006722173.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1800 | XP_006722236.1 | |
XM_006722174.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1793 | XP_006722237.1 | |
XM_006722175.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1740 | XP_006722238.1 | |
XM_006722176.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1523 | XP_006722239.1 | |
XM_006722177.3 | 5255 | Silent Mutation | GTA,GTG | V,V 1360 | XP_006722240.1 | |
XM_006722178.3 | 5255 | Silent Mutation | GTA,GTG | V,V 1360 | XP_006722241.1 | |
XM_011525471.2 | 5255 | Silent Mutation | GTA,GTG | V,V 1321 | XP_011523773.1 | |
XM_011525472.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1304 | XP_011523774.1 | |
XM_017025326.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1800 | XP_016880815.1 | |
XM_017025327.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1761 | XP_016880816.1 | |
XM_017025328.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1697 | XP_016880817.1 | |
XM_017025329.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1520 | XP_016880818.1 | |
XM_017025330.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1520 | XP_016880819.1 | |
XM_017025331.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1511 | XP_016880820.1 | |
XM_017025332.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1460 | XP_016880821.1 | |
XM_017025333.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1234 | XP_016880822.1 | |
XM_017025334.1 | 5255 | Silent Mutation | GTA,GTG | V,V 1183 | XP_016880823.1 |