Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGCCCTCTGGCGCTACCATGGC[A/G]TTTGGCAAGAGTCACCGGGATCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604701 | ||||||||||||||||||||
Literature Links: |
TOM1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOM1L1 - target of myb1 like 1 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321173.1 | 65 | Silent Mutation | GCA,GCG | A,A 2 | NP_001308102.1 | |
NM_001321174.1 | 65 | UTR 5 | NP_001308103.1 | |||
NM_001321175.1 | 65 | UTR 5 | NP_001308104.1 | |||
NM_001321176.1 | 65 | UTR 5 | NP_001308105.1 | |||
NM_005486.2 | 65 | Silent Mutation | GCA,GCG | A,A 2 | NP_005477.2 | |
XM_017024002.1 | 65 | UTR 5 | XP_016879491.1 |