Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAACACTTACCAGAGATGATGCC[A/G]ATGGCGACGTACATATGAGAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610008 MIM: 603880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A6 - solute carrier family 16 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174166.1 | 676 | Silent Mutation | ATC,ATT | I,I 121 | NP_001167637.1 | |
NM_004694.4 | 676 | Silent Mutation | ATC,ATT | I,I 121 | NP_004685.2 | |
XM_005257789.3 | 676 | Silent Mutation | ATC,ATT | I,I 125 | XP_005257846.2 | |
XM_011525461.2 | 676 | Silent Mutation | ATC,ATT | I,I 121 | XP_011523763.1 | |
XM_017025291.1 | 676 | Silent Mutation | ATC,ATT | I,I 121 | XP_016880780.1 | |
XM_017025292.1 | 676 | Silent Mutation | ATC,ATT | I,I 121 | XP_016880781.1 |