Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCTGCAGGGCCACGGCCTGCTG[C/G]GTCAGGCACTTGAGGTACTCGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616184 MIM: 601545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLUH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLUH - clustered mitochondria homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015229.3 | 3611 | Silent Mutation | ACC,ACG | T,T 1198 | NP_056044.3 | |
XM_011523768.2 | 3611 | Silent Mutation | ACC,ACG | T,T 1199 | XP_011522070.1 |
MIR6776 - microRNA 6776 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAFAH1B1 - platelet activating factor acetylhydrolase 1b regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |