Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTTGGAGGGGAAAACGCAATC[G/T]GATTTACCCTTGGCCGAGGTCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605829 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf99 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf99 - chromosome 17 open reading frame 99 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163075.1 | Intron | NP_001156547.1 | ||||
XM_011524152.2 | Intron | XP_011522454.1 | ||||
XM_017023997.1 | Intron | XP_016879486.1 | ||||
XM_017023998.1 | Intron | XP_016879487.1 | ||||
XM_017023999.1 | Intron | XP_016879488.1 | ||||
XM_017024000.1 | Intron | XP_016879489.1 | ||||
XM_017024001.1 | Intron | XP_016879490.1 |
TMC8 - transmembrane channel like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |