Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTAACATAGTAAGACATTGTA[A/G]AGGTGGAGGAGGAAGAAGGAATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929185 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929185 - uncharacterized LOC101929185 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VEZF1 - vascular endothelial zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007146.2 | Intron | NP_009077.2 | ||||
XM_005257643.1 | Intron | XP_005257700.1 | ||||
XM_005257644.4 | Intron | XP_005257701.1 | ||||
XM_017025017.1 | Intron | XP_016880506.1 | ||||
XM_017025018.1 | Intron | XP_016880507.1 |