Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGGATTTTCTGAGATCAGACACA[C/G]TTGGGCCGGGTTGGGAGGAACTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST3B1 - heparan sulfate-glucosamine 3-sulfotransferase 3B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006041.2 | Intron | NP_006032.1 | ||||
XM_017025479.1 | Intron | XP_016880968.1 |
MGC12916 - uncharacterized protein MGC12916 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |