Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCAACAGCAGCTACCAGAACA[A/C]CATCACGCACCTCAACAATGAGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603460 MIM: 606539 MIM: 604449 | ||||||||||||||||||||
Literature Links: |
CDK5R1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK5R1 - cyclin dependent kinase 5 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003885.2 | 438 | Missense Mutation | AAC,ACC | N,T 77 | NP_003876.1 | |
XM_011525438.2 | 438 | Missense Mutation | AAC,ACC | N,T 77 | XP_011523740.1 | |
XM_017025281.1 | 438 | Missense Mutation | AAC,ACC | N,T 77 | XP_016880770.1 |
MYO1D - myosin ID | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD11 - proteasome 26S subunit, non-ATPase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |