Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGGGGTGGGGCCCACTCGCC[A/G]GCCAGCAGCCCCTGTGTGGCCTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611825 MIM: 614966 MIM: 608613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC46 - leucine rich repeat containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033413.3 | 1185 | Intron | NP_219481.1 | |||
XM_005257775.4 | 1185 | Intron | XP_005257832.1 |
MRPL10 - mitochondrial ribosomal protein L10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN2 - secernin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145023.1 | 1185 | UTR 3 | NP_001138495.1 | |||
NM_138355.3 | 1185 | Silent Mutation | GCC,GCT | A,A 399 | NP_612364.2 | |
XM_005257776.4 | 1185 | Silent Mutation | GCC,GCT | A,A 399 | XP_005257833.1 | |
XM_017025286.1 | 1185 | Silent Mutation | GCC,GCT | A,A 377 | XP_016880775.1 |
SP6 - Sp6 transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |