Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTCCAGCAGCACCTGATACTCC[C/T]GGTTCTGCCGCTCCAGGTCACTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602761 | ||||||||||||||||||||
Literature Links: |
KRT33A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT33A - keratin 33A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004138.3 | 814 | Missense Mutation | CAG,CGG | Q,R 337 | NP_004129.2 | |
XM_011524786.2 | 814 | Missense Mutation | CAG,CGG | Q,R 178 | XP_011523088.1 |