Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGCAAAGAGTCCCTTGGTGCC[C/T]GACTCGGCCCGGATCCTCCGCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605448 MIM: 610820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUNDC3A - RUN domain containing 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A39 - solute carrier family 25 member 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143780.2 | 1032 | Silent Mutation | NP_001137252.1 | |||
NM_001321240.1 | 1032 | Silent Mutation | NP_001308169.1 | |||
NM_001321241.1 | 1032 | Silent Mutation | NP_001308170.1 | |||
NM_016016.3 | 1032 | Silent Mutation | NP_057100.1 | |||
XM_011524880.2 | 1032 | Silent Mutation | XP_011523182.1 | |||
XM_011524882.2 | 1032 | Silent Mutation | XP_011523184.1 | |||
XM_011524883.2 | 1032 | Silent Mutation | XP_011523185.1 | |||
XM_011524884.1 | 1032 | Silent Mutation | XP_011523186.1 | |||
XM_017024737.1 | 1032 | Silent Mutation | XP_016880226.1 |